Tisk…

Anti-STC1 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42105_P050EA 11140 CZK
AVIVARP42105_P050
Anti-STC1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: STC1 (stanniocalcin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR