Tisk…

Anti-LEFTY2 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42108_P050EA 10840 CZK
AVIVARP42108_P050
Anti-LEFTY2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LEFTY2 (left-right determination factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR