Tisk…

Anti-VGF Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42127_P050EA 11140 CZK
AVIVARP42127_P050
Anti-VGF Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: VGF (VGF nerve growth factor inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR