Tisk…

Anti-PPAP2A Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42145_P050EA 11140 CZK
AVIVARP42145_P050
Anti-PPAP2A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PPAP2A (phosphatidic acid phosphatase type 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR