Tisk…

Anti-CES2 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42158_P050EA 11140 CZK
AVIVARP42158_P050
Anti-CES2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CES2 (carboxylesterase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR