Tisk…

Anti-NUMB Rabbit Polyclonal Antibody [clone: 304CT11.6.1]

Dodavatel: Abgent
ABGEAP11449AEA 9810 CZK
ABGEAP11449A
Anti-NUMB Rabbit Polyclonal Antibody [clone: 304CT11.6.1]
Protilátky
Type: Primary
Antigen: NUMB (numb homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR