Tisk…

Anti-TMPRSS15 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP12255BEA 9870 CZK
ABGEAP12255B
Anti-TMPRSS15 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TMPRSS15 (transmembrane protease, serine 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR