Tisk…

Anti-EIF3G Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP5342CEA 9810 CZK
ABGEAP5342C
Anti-EIF3G Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EIF3G (eukaryotic translation initiation factor 3, subunit G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR