Tisk…

Anti-CDK1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP7517DEA 9870 CZK
ABGEAP7517D
Anti-CDK1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR