Tisk…

Anti-TRPM6 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP8053CEA 9810 CZK
ABGEAP8053C
Anti-TRPM6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRPM6 (transient receptor potential cation channel, subfamily M, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR