Tisk…

Anti-UNC13B Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP8905CEA 10150 CZK
ABGEAP8905C
Anti-UNC13B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: UNC13B (unc-13 homolog B (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR