Tisk…
Anti-GRIA1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2261AEA 9870 CZK
ABGEAT2261A
Anti-GRIA1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: GRIA1 (glutamate receptor, ionotropic, AMPA 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human