Tisk…
Anti-GRID2 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2263AEA 9870 CZK
ABGEAT2263A
Anti-GRID2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: GRID2 (glutamate receptor, ionotropic, delta 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human