Tisk…

Anti-GUCY2C Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2299AEA 9870 CZK
ABGEAT2299A
Anti-GUCY2C Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: GUCY2C (guanylate cyclase 2C (heat stable enterotoxin receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR