Tisk…

Anti-HSPB7 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2449AEA 9870 CZK
ABGEAT2449A
Anti-HSPB7 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HSPB7 (heat shock 27kDa protein family, member 7 (cardiovascular))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR