Tisk…
Anti-HTR5A Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2457AEA 9870 CZK
ABGEAT2457A
Anti-HTR5A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HTR5A (5-hydroxytryptamine (serotonin) receptor 5A, G protein-coupled)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human