Tisk…
Anti-IFNE Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2489AEA 9870 CZK
ABGEAT2489A
Anti-IFNE Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IFNE (interferon, epsilon)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human