Tisk…
Anti-IFRD2 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2490AEA 9870 CZK
ABGEAT2490A
Anti-IFRD2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IFRD2 (interferon-related developmental regulator 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human