Tisk…
Anti-IFT57 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2491AEA 9870 CZK
ABGEAT2491A
Anti-IFT57 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IFT57 (intraflagellar transport 57 homolog (Chlamydomonas))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human