Tisk…

Anti-IL17B Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2506AEA 9870 CZK
ABGEAT2506A
Anti-IL17B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IL17B (interleukin 17B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR