Tisk…

Anti-IL6ST Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2524AEA 9870 CZK
ABGEAT2524A
Anti-IL6ST Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IL6ST (interleukin 6 signal transducer (gp130, oncostatin M receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR