Tisk…
Anti-IRF9 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2558AEA 9870 CZK
ABGEAT2558A
Anti-IRF9 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IRF9 (interferon regulatory factor 9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human