Tisk…

Anti-ISL1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2560AEA 9870 CZK
ABGEAT2560A
Anti-ISL1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ISL1 (ISL LIM homeobox 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR