Tisk…

Anti-ITGB2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2568AEA 9870 CZK
ABGEAT2568A
Anti-ITGB2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR