Tisk…

Anti-STT3A Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2572AEA 9870 CZK
ABGEAT2572A
Anti-STT3A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: STT3A (STT3A, subunit of the oligosaccharyltransferase complex (catalytic))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR