Tisk…
Anti-JAM3 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2579AEA 9870 CZK
ABGEAT2579A
Anti-JAM3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: JAM3 (junctional adhesion molecule 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human