Tisk…

Anti-KDM5A Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2580AEA 9870 CZK
ABGEAT2580A
Anti-KDM5A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: KDM5A (lysine (K)-specific demethylase 5A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR