Tisk…

Anti-LDHAL6B Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2688AEA 9870 CZK
ABGEAT2688A
Anti-LDHAL6B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: LDHAL6B (lactate dehydrogenase A-like 6B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR