Tisk…
Anti-LILRB1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2715AEA 9870 CZK
ABGEAT2715A
Anti-LILRB1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: LILRB1 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human