Tisk…
Anti-LRRFIP1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2740AEA 9870 CZK
ABGEAT2740A
Anti-LRRFIP1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: LRRFIP1 (leucine rich repeat (in FLII) interacting protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human