Tisk…

Anti-LRRFIP1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2741AEA 9870 CZK
ABGEAT2741A
Anti-LRRFIP1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: LRRFIP1 (leucine rich repeat (in FLII) interacting protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR