Tisk…

Anti-LY75 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2752AEA 9870 CZK
ABGEAT2752A
Anti-LY75 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: LY75 (lymphocyte antigen 75)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR