Tisk…
Anti-MAP2K5 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2772AEA 9870 CZK
ABGEAT2772A
Anti-MAP2K5 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MAP2K5 (mitogen-activated protein kinase kinase 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human