Tisk…

Anti-MAP3K5 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2777AEA 9870 CZK
ABGEAT2777A
Anti-MAP3K5 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MAP3K5 (mitogen-activated protein kinase kinase kinase 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR