Tisk…

Anti-MARCH7 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2801AEA 9870 CZK
ABGEAT2801A
Anti-MARCH7 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MARCH7 (membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR