Tisk…

Anti-MAZ Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2809AEA 9870 CZK
ABGEAT2809A
Anti-MAZ Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MAZ (MYC-associated zinc finger protein (purine-binding transcription factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR