Tisk…

Anti-MGAT4A Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2855AEA 9870 CZK
ABGEAT2855A
Anti-MGAT4A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MGAT4A (mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR