Tisk…

Anti-MIB2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2868AEA 9870 CZK
ABGEAT2868A
Anti-MIB2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MIB2 (mindbomb E3 ubiquitin protein ligase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR