Tisk…

Anti-MYL6B Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2877AEA 9870 CZK
ABGEAT2877A
Anti-MYL6B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MYL6B (myosin, light chain 6B, alkali, smooth muscle and non-muscle)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR