Tisk…

Anti-MRE11A Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2901AEA 9870 CZK
ABGEAT2901A
Anti-MRE11A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MRE11A (MRE11 meiotic recombination 11 homolog A (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR