Tisk…
Anti-MYOG Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT2964AEA 9870 CZK
ABGEAT2964A
Anti-MYOG Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MYOG (myogenin (myogenic factor 4))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human