Tisk…

Anti-NDN Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT2988AEA 9870 CZK
ABGEAT2988A
Anti-NDN Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NDN (necdin, melanoma antigen (MAGE) family member)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR