Tisk…

Anti-NDUFB11 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3001AEA 9870 CZK
ABGEAT3001A
Anti-NDUFB11 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NDUFB11 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR