Tisk…

Anti-NDUFB5 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3004AEA 9870 CZK
ABGEAT3004A
Anti-NDUFB5 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NDUFB5 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR