Tisk…

Anti-NDUFS4 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3007AEA 9870 CZK
ABGEAT3007A
Anti-NDUFS4 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NDUFS4 (NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR