Tisk…

Anti-NEIL2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3012AEA 9870 CZK
ABGEAT3012A
Anti-NEIL2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NEIL2 (nei endonuclease VIII-like 2 (E. coli))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR