Tisk…

Anti-NEK11 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3014AEA 9870 CZK
ABGEAT3014A
Anti-NEK11 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NEK11 (NIMA-related kinase 11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR