Tisk…
Anti-NFIC Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3033AEA 9870 CZK
ABGEAT3033A
Anti-NFIC Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NFIC (nuclear factor I/C (CCAAT-binding transcription factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human