Tisk…

Anti-NR0B1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3091AEA 9870 CZK
ABGEAT3091A
Anti-NR0B1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NR0B1 (nuclear receptor subfamily 0, group B, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR