Tisk…

Anti-NR1D1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3093AEA 9870 CZK
ABGEAT3093A
Anti-NR1D1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: NR1D1 (nuclear receptor subfamily 1, group D, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR